4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) überwiegend trans ≥98%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: 4-(tert-butyl)cyclohexanecarboxylic acid
L08248.06 L08248.14
L08248.06EA 47.9 EUR
L08248.06 L08248.14
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture) überwiegend trans ≥98%
4-tert-Butylcyclohexanecarboxylic acid (cis and trans mixture)
Purity: 98+% CAS No.: 5451-55-8 Molecular Formula: C11H20O2 Formula Weight: 184.28 Beilstein Reference: 2043187 MDL No.: MFCD00042622
Formel: C₁₁H₂₀O₂
Molecular Weight: 184.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00042622
CAS-Nummer: 5451-55-8