N6-Benzoyladenine 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Benzamidopurine, N-(7H-Purin-6-yl)benzamide
L08292.03
L08292.06
L08292.14
L08292.03EA
43.6
EUR
L08292.03
L08292.06
L08292.14
N6-Benzoyladenine 99%
N6-Benzoyladenine
Purity: 99%
CAS No.: 4005-49-6
Molecular Formula: C12H9N5O
Formula Weight: 239.23
Melting Point: 241-244°
Beilstein Reference: 20585
MDL No.: MFCD00037927
Formel:
C₁₂H₉N₅O Molecular Weight: 239,24 g/mol Schmelzpunkt: 241…244 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037927 CAS-Nummer: 4005-49-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...