5-Cyanouracil 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Uracil-5-carbonitrile, 2,4-Dihydroxypyrimidine-5-carbonitrile, 2,4-Dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile, 5-Cyano-2,4-dioxo-1,2,3,4-tetrahydropyrimidine

L08490.03 L08490.06
L08490.03EA 103 EUR
L08490.03 L08490.06
5-Cyanouracil 97%
5-Cyanouracil

Purity: 97% CAS No.: 5428-41-1 Molecular Formula: C5H3N3O2 Formula Weight: 137.10 Melting Point: 300° dec. Beilstein Reference: 5707 UN No.: UN3439 MDL No.: MFCD00051958

Formel: C₅H₃N₃O₂
Molecular Weight: 137,10 g/mol
Schmelzpunkt: >300 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00051958
CAS-Nummer: 5428-41-1
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR