Dimethyl-5-norbornene-2,3-dicarboxylate 94%
Lieferant: Thermo Scientific
|
Achtung
|
L08664.09
L08664.18
L08664.09EA
80.2
EUR
L08664.09
L08664.18
Dimethyl-5-norbornene-2,3-dicarboxylate 94%
Dimethyl-5-norbornene-2,3-dicarboxylate
Purity: 94%
CAS No.: 5826-73-3
Molecular Formula: C11H14O4
Formula Weight: 210.23
Boiling Point: 150-152°/20mm
Beilstein Reference: 1248588
MDL No.: MFCD00154455
Formel:
C₁₁H₁₄O₄ Molecular Weight: 210,23 g/mol Siedepunkt: 150…152 °C (20 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00154455 CAS-Nummer: 5826-73-3 Merck Index: 14,3240 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...