3,4-(Methylenedioxy)benzylideneacetone 98%

Lieferant: Thermo Scientific
Achtung

L08675.09 L08675.18
L08675.09EA 57.7 EUR
L08675.09 L08675.18
3,4-(Methylenedioxy)benzylideneacetone 98%
3,4-(Methylenedioxy)benzylideneacetone

Purity: 98% CAS No.: 3160-37-0 Molecular Formula: C11H10O3 Formula Weight: 190.20 Melting Point: 108-110° Beilstein Reference: 83983 MDL No.: MFCD00016907

Formel: C₁₁H₁₀O₃
Molecular Weight: 190,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016907
CAS-Nummer: 3160-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR