Ethyl indole-3-acrylate 97%
Lieferant: Thermo Scientific
L08703.03
L08703.06
L08703.03EA
43.5
EUR
L08703.03
L08703.06
Ethyl indole-3-acrylate 97%
Ethyl indole-3-acrylate
Purity: 97%
CAS No.: 15181-86-9
Molecular Formula: C13H13NO2
Formula Weight: 215.25
Melting Point: 118-122°
Beilstein Reference: 476317
MDL No.: MFCD00051947
Formel:
C₁₃H₁₃NO₂ Molecular Weight: 215,25 g/mol Schmelzpunkt: 118…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051947 CAS-Nummer: 15181-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...