N-(tert-Butoxycarbonyl)-D-phenylalanine 98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Boc-D-phenylalanine, BOC-D-Phenylalanine, Boc-D-Phe-OH

L08722.03 L08722.06
L08722.03EA 26.6 EUR
L08722.03 L08722.06
N-(tert-Butoxycarbonyl)-D-phenylalanine 98%
N-(tert-Butoxycarbonyl)-D-phenylalanine

Purity: 98% CAS No.: 18942-49-9 Molecular Formula: C14H19NO4 Formula Weight: 265.31 Storage Temperature: 2°C to 8°C Melting Point: 85-88° Beilstein Reference: 3593396 Optical Rotation: -24.5° (c=1 in ethanol) MDL No.: MFCD00063149

Formel: C₁₄H₁₉NO₄
Schmelzpunkt: 85…88 °C
MDL: MFCD00063149
CAS-Nummer: 18942-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR