Cbz-D-Valin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Cbz-D-Val-OH, Z-D-Val-OH, N-Cbz-D-valine

L08856.03 L08856.06
L08856.03EA 80.3 EUR
L08856.03 L08856.06
Cbz-D-Valin ≥98%
Cbz-D-Valin

Purity: 98+% CAS No.: 1685-33-2 Molecular Formula: C13H17NO4 Formula Weight: 251.28 Storage Temperature: 2°C to 8°C Melting Point: 58-60° Beilstein Reference: 2056616 Optical Rotation: +3.8° (c=2 in acetic acid) MDL No.: MFCD00065703

Formel: C₁₃H₁₇NO₄
Molecular Weight: 251,28 g/mol
Schmelzpunkt: 58…60 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00065703
CAS-Nummer: 1685-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR