1,2-Di(p-tolyl)ethane 98%

Lieferant: Thermo Scientific
Achtung

L08876.03 L08876.06
L08876.03EA 36 EUR
L08876.03 L08876.06
1,2-Di(p-tolyl)ethane 98%
1,2-Di(p-tolyl)ethane

Purity: 98% CAS No.: 538-39-6 Molecular Formula: C16H18 Formula Weight: 210.32 Melting Point: 80-82° Boiling Point: 296-298°/730mm Beilstein Reference: 1909630 MDL No.: MFCD00026025

Formel: C₁₆H₁₈
Molecular Weight: 210,32 g/mol
Siedepunkt: 296…298 °C (730 mmHg)
Schmelzpunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00026025
CAS-Nummer: 538-39-6
Merck Index: 14,3380

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR