4-Benzyl-L-aspartate 98%

Lieferant: Thermo Fisher Scientific

Synonyme: L-Aspartic acid beta-benzyl ester, β-Benzyl L-aspartate, H-Asp(obzl)-OH, beta-Benzyl L-aspartate, L-Aspartic acid 4-benzyl ester, L-Aspartic Acid 4-Benzyl Ester, beta-Benzyl L-Aspartate, H-Asp(OBzl)-OH, 4-Benzyl L-Aspartate, H-Asp(OBn)-OH, L-Aspartic Acid beta-Benzyl Ester, L-Aspartic acid β-benzyl ester

L08956.06 L08956.14
L08956.06EA 71.9 EUR
L08956.06 L08956.14
4-Benzyl-L-aspartate 98%
4-Benzyl-L-aspartate

Purity: 98% CAS No.: 2177-63-1 Molecular Formula: C11H13NO4 Formula Weight: 223.23 Melting Point: ca 220° dec. Beilstein Reference: 1983183 Optical Rotation: +27.5° (c=1 in 1N HCl) MDL No.: MFCD00037208

Formel: C₁₁H₁₃NO₄
Molecular Weight: 223.23 g/mol
Schmelzpunkt: ∼ 220 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00037208
CAS-Nummer: 2177-63-1
EINECS: 218-541-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR