N-Benzyloxycarbonyl-6-Aminohexansäure 99%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Cbz-epsilon-aminocaproic Acid, N-Cbz-6-aminohexanoic Acid, N-Benzyloxycarbonyl-6-aminohexanoic acid, N-Carbobenzoxy-epsilon-aminocaproic Acid, Z-ε-Acp-OH

L08997.06 L08997.14
L08997.06EA 43.5 EUR
L08997.06 L08997.14
N-Benzyloxycarbonyl-6-Aminohexansäure 99%
N-Benzyloxycarbonyl-6-Aminohexansäure

Purity: 99% CAS No.: 1947-00-8 Molecular Formula: C6H5CH2OCONH(CH2)5CO2H Formula Weight: 265.31 Storage Temperature: 2°C to 8°C Melting Point: 53-57° Beilstein Reference: 2288508 MDL No.: MFCD00004423

Formel: C₁₄H₁₉NO₄
Molecular Weight: 265.31 g/mol
Schmelzpunkt: 53…57 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00004423
CAS-Nummer: 1947-00-8
EINECS: 217-748-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR