3,3',5,5'-Tetramethylbiphenyl ≥97%

Lieferant: Thermo Scientific

Synonyme: 3,3',5,5'-Tetramethyl-1,1'-biphenyl

L09172.03 L09172.06
L09172.03EA 60.1 EUR
L09172.03 L09172.06
3,3',5,5'-Tetramethylbiphenyl ≥97%
3,3',5,5'-Tetramethylbiphenyl

Purity: 97+% CAS No.: 25570-02-9 Molecular Formula: C16H18 Formula Weight: 210.32 Melting Point: 47-49° Beilstein Reference: 1935931 MDL No.: MFCD00051910

Formel: C₁₆H₁₈
Molecular Weight: 210,32 g/mol
Schmelzpunkt: 47…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051910
CAS-Nummer: 25570-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR