N-Vinylphthalimid 99%

Lieferant: Thermo Fisher Scientific
Achtung

L09335.03 L09335.06
L09335.03EA 37.6 EUR
L09335.03 L09335.06
N-Vinylphthalimid 99%
N-Vinylphthalimid

Purity: 99% CAS No.: 3485-84-5 Molecular Formula: C10H7NO2 Formula Weight: 173.17 Odour: Odourless Melting Point: 84-88°Beilstein Reference: 127880 MDL No.: MFCD00078446

Formel: C₁₀H₇NO₂
Molecular Weight: 173.17 g/mol
Schmelzpunkt: 84…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078446
CAS-Nummer: 3485-84-5
EINECS: 222-473-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR