(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 95%
Lieferant: Thermo Scientific
Synonyme:
H-Lys(Z)-OMe.HCl, H-Lys(Z)-OMe·HCl
L09744.03
L09744.06
L09744.03EA
33.2
EUR
L09744.03
L09744.06
(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride 95%
(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Purity: 95%
CAS No.: 27894-50-4
Molecular Formula: C15H22N2O4.HCl
Formula Weight: 330.81
Storage Temperature: 2°C to 8°C
Melting Point: 115-118°
Beilstein Reference: 3578476
Optical Rotation: +14.5° (c=2 in water)
MDL No.: MFCD00034846
Formel:
C₁₅H₂₃ClN₂O₄ Molecular Weight: 330,809 g/mol Schmelzpunkt: 115…118 °C |
MDL:
MFCD00034846 CAS-Nummer: 27894-50-4 EINECS: 248-715-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...