Zinc-copper couple (∼1 - 3% Cu)
Lieferant: Thermo Scientific
|
Achtung
|
L09811.14
L09811.22
L09811.14EA
72.5
EUR
L09811.14
L09811.22
Zinc-copper couple (∼1 - 3% Cu)
Zinc-copper couple
CAS No.: 53801-63-1
Molecular Formula: Zn/Cu
Storage Temperature: 2°C to 8°C
Melting Point: 419-420°
UN No.: UN3089
MDL No.: MFCD00084851
Formel:
CuZn Molecular Weight: 128,926 g/mol Schmelzpunkt: 419…420 °C |
MDL:
MFCD00084851 CAS-Nummer: 53801-63-1 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...