Phenyl trifluoromethyl sulphide 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Phenyl(trifluoromethyl)sulphane, alpha,alpha,alpha-Trifluorothioanisole, (Trifluoromethylthio)benzene
L10114.03
L10114.06
L10114.03EA
30.2
EUR
L10114.03
L10114.06
Phenyl trifluoromethyl sulphide 98%
Phenyl trifluoromethyl sulphide
Purity: 98%
CAS No.: 456-56-4
Molecular Formula: C7H5F3S
Formula Weight: 178.18
Flash Point: 37°(98°F)
Refractive Index: 1.4655
Odour: Unpleasant
Boiling Point: 140-142°
Beilstein Reference: 2043134
UN No.: UN1993
MDL No.: MFCD00040839
Formel:
C₇H₅F₃S Molecular Weight: 178,17 g/mol Siedepunkt: 140…142 °C Dichte: 1,249 Flammpunkt: 37 °C (98 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040839 CAS-Nummer: 456-56-4 EINECS: 207-270-0 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...