2-[(2-Methylphenoxy)methyl]oxirane ≥85%, tech.

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-((o-Tolyloxy)methyl)oxirane

L10136.14 L10136.22
L10136.14EA 28.3 EUR
L10136.14 L10136.22
2-[(2-Methylphenoxy)methyl]oxirane ≥85%, tech.
2-[(2-Methylphenoxy)methyl]oxirane

Purity: 85% CAS No.: 2210-79-9 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 120°(248°F) Refractive Index: 1.5290 Boiling Point: 119-120°/6mm Beilstein Reference: 4585 UN No.: UN3082 MDL No.: MFCD00037847

Formel: C₁₀H₁₂O₂
Molecular Weight: 164,21 g/mol
Siedepunkt: 119…120 °C (6 mmHg)
Dichte: 1,079
Flammpunkt: 120 °C (248 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00037847
CAS-Nummer: 2210-79-9
EINECS: 218-645-3
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR