Naringin hydrate 98%

Lieferant: Thermo Scientific

Synonyme: (S)-7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-2-(4-hydroxyphenyl)chroman-4-one hydrate, Naringenin 7-Rhamnoglucoside hydrate

L10163.09 L10163.18
L10163.09EA 25.2 EUR
L10163.09 L10163.18
Naringin hydrate 98%
Naringin hydrate

Purity: 98% CAS No.: 10236-47-2 Molecular Formula: C27H32O14.xH2O Formula Weight: 580.55(anhy) Melting Point: ca 166° dec. Beilstein Reference: 102012 Optical Rotation: -90° (c=1 in ethanol) MDL No.: MFCD00149445

Formel: C₂₇H₃₂O₁₄
Molecular Weight: 598,55 g/mol
Schmelzpunkt: ∼ 166 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00148888
CAS-Nummer: 10236-47-2
EINECS: 233-566-4
Merck Index: 14,6424

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR