2,2,3,3,3-Pentafluoropropyl acrylate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 356-86-5
Molecular Formula: C6H5F5O2
Formula Weight: 204.10
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Refractive Index: 1.3360
Odour: Pungent
Boiling Point: 49-50°/100mm
Beilstein Reference: 1779140
UN No.: UN3272
MDL No.: MFCD00039257
Formel:
C₆H₅F₅O₂ Siedepunkt: 49…50 °C (100 mmHg) Dichte: 1,320 Flammpunkt: 23 °C (73 °F) |
MDL:
MFCD00039257 CAS-Nummer: 356-86-5 EINECS: 206-608-4 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...