(2E)-4-(4-Methylphenyl)-4-oxobut-2-enoic acid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(4-Methylbenzoyl)acrylic acid
L10551.06
L10551.03
L10551.06EA
80.5
EUR
L10551.06
L10551.03
(2E)-4-(4-Methylphenyl)-4-oxobut-2-enoic acid 98%
(2E)-4-(4-Methylphenyl)-4-oxobut-2-enoic acid
Purity: 98%
CAS No.: 20972-36-5
Molecular Formula: C11H10O3
Formula Weight: 190.20
Melting Point: 138-140°
Beilstein Reference: 2691231
MDL No.: MFCD00075338
Formel:
C₁₁H₁₀O₃ Molecular Weight: 190,2 g/mol Schmelzpunkt: 138…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075338 CAS-Nummer: 20972-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...