4-Amino-1,3,5-trimethyl-1H-pyrazole 97%
Lieferant: Thermo Scientific
Synonyme:
1,3,5-Trimethyl-1H-pyrazol-4-amine
L10625.03
L10625.06
L10625.03EA
71.3
EUR
L10625.03
L10625.06
4-Amino-1,3,5-trimethyl-1H-pyrazole 97%
4-Amino-1,3,5-trimethyl-1H-pyrazole
Purity: 97%
CAS No.: 28466-21-9
Molecular Formula: C6H11N3
Formula Weight: 125.17
Melting Point: 99-105°
Beilstein Reference: 110739
MDL No.: MFCD00052883
Formel:
C₆H₁₁N₃ Molecular Weight: 125,17 g/mol Schmelzpunkt: 99…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052883 CAS-Nummer: 28466-21-9 |