Methyl thiazolidine-2-carboxylate hydrochloride 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Thiazolidine-2-carboxylic acid methyl ester hydrochloride
L10740.03
L10740.06
L10740.03EA
28.4
EUR
L10740.03
L10740.06
Methyl thiazolidine-2-carboxylate hydrochloride 98%
Methyl thiazolidine-2-carboxylate hydrochloride
Purity: 98%
CAS No.: 50703-06-5
Molecular Formula: C5H9NO2S.HCl
Formula Weight: 183.66
Melting Point: ca 158° dec.
Beilstein Reference: 3628640
MDL No.: MFCD00068022
Formel:
C₅H₉NO₂S·HCl Molecular Weight: 183.656 g/mol Schmelzpunkt: ∼ 158 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068022 CAS-Nummer: 50703-06-5 EINECS: 256-726-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...