DL-Alanine ethyl ester hydrochloride 99%
Lieferant: Thermo Scientific
Synonyme: Ethyl 2-aminopropanoate hydrochloride, H-DL-Ala-OEt.HCl
L11020.06 L11020.14
L11020.06EA 46.1 EUR
L11020.06 L11020.14
DL-Alanine ethyl ester hydrochloride 99%
DL-Alanine ethyl ester hydrochloride
Purity: 99% CAS No.: 617-27-6 Molecular Formula: C5H11NO2.HCl Formula Weight: 153.61 Melting Point: 84-86° Beilstein Reference: 3654425 MDL No.: MFCD00013018
Formel: C₅H₁₂ClNO₂
Molecular Weight: 153,606 g/mol
Schmelzpunkt: 84…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013018
CAS-Nummer: 617-27-6
EINECS: 210-507-0