N-Acetyl-D-mannosamine monohydrate 99%
Lieferant: Thermo Fisher Scientific
L11167.03
L11167.06
L11167.03EA
58.3
EUR
L11167.03
L11167.06
N-Acetyl-D-mannosamine monohydrate 99%
N-Acetyl-D-mannosamine monohydrate
Purity: 99%
CAS No.: 7772-94-3
Molecular Formula: C8H15NO6.H2O
Formula Weight: 239.24 (221.22anhy)
Storage Temperature: 2°C to 8°C
Melting Point: ca 130° dec.
Beilstein Reference: 1346524
Optical Rotation: +10.5° (c=1 in water)
MDL No.: MFCD00149493
Formel:
C₈H₁₅NO₆·H₂O Schmelzpunkt: ∼ 130 °C (Zersetzung) |
MDL:
MFCD00149493 CAS-Nummer: 7772-94-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...