Benzo[b]thiophene-2-carbohydrazide 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Benzo[b]thiophene-2-carboxylic hydrazide

L11357.03 L11357.06
L11357.03EA 64.8 EUR
L11357.03 L11357.06
Benzo[b]thiophene-2-carbohydrazide 97%
Benzo[b]thiophene-2-carbohydrazide

Purity: 97% CAS No.: 175135-07-6 Molecular Formula: C9H8N2OS Formula Weight: 192.24 Melting Point: 184-187° Beilstein Reference: 153297 MDL No.: MFCD00052501

Formel: C₉H₈N₂OS
Molecular Weight: 192.236 g/mol
Schmelzpunkt: 184…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052501
CAS-Nummer: 175135-07-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR