3-(4-Chlorobenzoyl)acrylic acid 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2E)-4-(4-Chlorophenyl)-4-oxobut-2-enoic acid

L11467.06 L11467.14
L11467.06EA 28.1 EUR
L11467.06 L11467.14
3-(4-Chlorobenzoyl)acrylic acid 99%
3-(4-Chlorobenzoyl)acrylic acid

Purity: 99% CAS No.: 6269-33-6 Molecular Formula: C10H7ClO3 Formula Weight: 210.62 Melting Point: 157-159° Beilstein Reference: 2719881 MDL No.: MFCD00041465

Formel: C₁₀H₇ClO₃
Molecular Weight: 210,62 g/mol
Schmelzpunkt: 157…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041465
CAS-Nummer: 6269-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR