Aminoguanidine nitrate, (max. 10% H₂O) 98%

Lieferant: Thermo Scientific
Gefahr

L11632.14 L11632.22
L11632.14EA 34.7 EUR
L11632.14 L11632.22
Aminoguanidine nitrate, (max. 10% H₂O) 98%
Aminoguanidine nitrate

Purity: 98% CAS No.: 10308-82-4 Molecular Formula: CH6N4.HNO3 Formula Weight: 137.10 Melting Point: 138-142° Beilstein Reference: 3916056 UN No.: UN1479 MDL No.: MFCD00013174

Formel: CH₇N₅O₃
Molecular Weight: 137,099 g/mol
Schmelzpunkt: 138…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013174
CAS-Nummer: 10308-82-4
EINECS: 233-682-5
UN: 1479
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR