Mandelic acid hydrazide 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-2-phenylacetohydrazide
L11653.06
L11653.14
L11653.06EA
64.6
EUR
L11653.06
L11653.14
Mandelic acid hydrazide 97%
Mandelic acid hydrazide
Purity: 97%
CAS No.: 2443-66-5
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Melting Point: 128-131°
Beilstein Reference: 2096197
MDL No.: MFCD00038133
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166,18 g/mol Schmelzpunkt: 128…131 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038133 CAS-Nummer: 2443-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...