Phenyl acrylate 97%
Lieferant: Thermo Scientific
|
Achtung
|
L11819.06
L11819.14
L11819.06EA
76.9
EUR
L11819.06
L11819.14
Phenyl acrylate 97%
Phenyl acrylate
Purity: 97%
CAS No.: 937-41-7
Molecular Formula: C6H5O2CCH=CH2
Formula Weight: 148.16
Refractive Index: 1.5210
Boiling Point: 89-90°/12mm
Beilstein Reference: 2041124
UN No.: UN3082
MDL No.: MFCD00048145
Formel:
C₉H₈O₂ Siedepunkt: 89…90 °C (12 mmHg) Dichte: 1,076 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048145 CAS-Nummer: 937-41-7 EINECS: 213-329-1 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...