Mono-n-dodecyl phosphate 90%, tech.
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dihydrogen monolauryl phosphate
L11941.06
L11941.14
L11941.06EA
119
EUR
L11941.06
L11941.14
Mono-n-dodecyl phosphate 90%, tech.
Mono-n-dodecyl phosphate
Purity: 90%
CAS No.: 2627-35-2
Molecular Formula: C12H27O4P
Formula Weight: 266.32
Odour: Odourless
Melting Point: 39-45°
Beilstein Reference: 1710518
MDL No.: MFCD00013946
Formel:
C₁₂H₂₇O₄P Molecular Weight: 266,318 g/mol Schmelzpunkt: 39…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013946 CAS-Nummer: 2627-35-2 EINECS: 220-095-4 |