4-Nitrophenyl octanoate 96%
Lieferant: Thermo Fisher Scientific
L12022.03
L12022.06
L12022.03EA
28.9
EUR
L12022.03
L12022.06
4-Nitrophenyl octanoate 96%
4-Nitrophenyl octanoate
Refractive Index: 1.5130
Formel:
C₁₄H₁₉NO₄ Dichte: 1,082 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024665 CAS-Nummer: 1956-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...