Thiophene-3-acetic acid hydrazide 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(Thien-3-yl)acetohydrazide

L12077.03 L12077.06
L12077.03EA 74.1 EUR
L12077.03 L12077.06
Thiophene-3-acetic acid hydrazide 97%
Thiophene-3-acetic acid hydrazide

Purity: 97% CAS No.: 175276-94-5 Molecular Formula: C6H8N2OS Formula Weight: 156.21 Melting Point: 85-86° Beilstein Reference: 120473 MDL No.: MFCD00052946

Formel: C₆H₈N₂OS
Molecular Weight: 156,21 g/mol
Schmelzpunkt: 85…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052946
CAS-Nummer: 175276-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR