(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
Lieferant: Thermo Scientific
Synonyme:
1,2:5,6-Di-O-isopropylidene-D-mannitol, (1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol, (1S,2S)-1,2-BIS((R)-2,2-DIMETHYL-1,3-DIOXOLAN-4-YL)ETHANE-1,2-DIOL
L12127.06
L12127.14
L12127.06EA
48.2
EUR
L12127.06
L12127.14
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
CAS No.: 1707-77-3
Molecular Formula: C12H22O6
Formula Weight: 262.31
Storage Temperature: 2°C to 8°C
Melting Point: 120-122°
Beilstein Reference: 83765
Optical Rotation: +6° (c=5 in chloroform)
MDL No.: MFCD00003211
Formel:
C₁₂H₂₂O₆ Molecular Weight: 262,302 g/mol Schmelzpunkt: 120…122 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00003211 CAS-Nummer: 1707-77-3 EINECS: 216-954-8 |