N-Benzoylthiourea 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-Carbamothioylbenzamide, N-(Carbamothioyl)benzamide
L12377.06
L12377.14
L12377.06EA
27.2
EUR
L12377.06
L12377.14
N-Benzoylthiourea 98%
N-Benzoylthiourea
Purity: 98%
CAS No.: 614-23-3
Molecular Formula: C8H8N2OS
Formula Weight: 180.23
Odour: Odourless
Melting Point: 174-176°
Beilstein Reference: 1942772
MDL No.: MFCD00041191
Formel:
C₈H₈N₂OS Molecular Weight: 180,23 g/mol Schmelzpunkt: 174…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041191 CAS-Nummer: 614-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...