2-Amino-4-hydroxyquinoline hydrate, (max. 10% H₂O) 97%

Lieferant: Thermo Fisher Scientific
Achtung

L12760.03 L12760.06
L12760.03EA 29.6 EUR
L12760.03 L12760.06
2-Amino-4-hydroxyquinoline hydrate, (max. 10% H₂O) 97%
2-Amino-4-hydroxyquinoline hydrate

Purity: 97% CAS No.: 42712-64-1 Molecular Formula: C9H8N2O.xH2O Formula Weight: 160.18(anhy) Melting Point: ca 298° dec. Beilstein Reference: 124785 MDL No.: MFCD00052386

Formel: C₉H₈N₂O·xH₂O
Molecular Weight: 160.18 g/mol (anhy)
Schmelzpunkt: ∼ 298 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052386
CAS-Nummer: 42712-64-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR