3-Ethylamino-p-cresol 90%, tech.

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Ethylamino-4-methylphenol

L12768.06 L12768.14
L12768.06EA 43.8 EUR
L12768.06 L12768.14
3-Ethylamino-p-cresol 90%, tech.
3-Ethylamino-p-cresol

Purity: 90% CAS No.: 120-37-6 Molecular Formula: C9H13NO Formula Weight: 151.21 Melting Point: 84-92° Boiling Point: 135-140°/3mm Beilstein Reference: 3241548 MDL No.: MFCD00002382

Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
Siedepunkt: 135…140 °C (3 mmHg)
Schmelzpunkt: 84…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002382
CAS-Nummer: 120-37-6
EINECS: 204-391-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR