Hydroxypivalic acid ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,2-Dimethyl-3-hydroxypropionic Acid, 3-Hydroxy-2,2-dimethylpropanoic acid, 3-Hydroxy-2,2-dimethylpropionic acid

L12858.18 L12858.30
L12858.18EA 76.3 EUR
L12858.18 L12858.30
Hydroxypivalic acid ≥97%
Hydroxypivalic acid

Purity: 98% CAS No.: 4835-90-9 Molecular Formula: C5H10O3 Formula Weight: 118.13 Odour: Odourless Melting Point: 122-126° Beilstein Reference: 1747025 MDL No.: MFCD00059953

Formel: C₅H₁₀O₃
Molecular Weight: 118,13 g/mol
Schmelzpunkt: 122…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00059953
CAS-Nummer: 4835-90-9
EINECS: 225-419-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR