Domiphen bromide 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (Dodecyldimethyl-2-phenoxyethyl)ammonium bromide, N,N-Dimethyl-N-(2-phenoxyethyl)dodecan-1-aminium bromide

L12930.14 L12930.22
L12930.14EA 69.6 EUR
L12930.14 L12930.22
Domiphen bromide 97%
Domiphen bromide

Purity: 97% CAS No.: 538-71-6 Molecular Formula: C22H40BrNO Formula Weight: 414.48 Melting Point: 117-118° Beilstein Reference: 3922742 MDL No.: MFCD00011769

Formel: C₂₂H₄₀BrNO
Molecular Weight: 414.48 g/mol
Schmelzpunkt: 117…118 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011769
CAS-Nummer: 538-71-6
EINECS: 208-702-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR