3-(Methoxycarbonyl)benzoesäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Isophthalic acid monomethyl ester, 3-(Methoxycarbonyl)benzoic acid, Benzene-1,3-dicarboxylic acid monomethyl ester, mono-Methyl isophthalate
L12992.03
L12992.06
L12992.03EA
19.1
EUR
L12992.03
L12992.06
3-(Methoxycarbonyl)benzoesäure 97%
Methyl hydrogen isophthalate
Purity: 97%
CAS No.: 1877-71-0
Molecular Formula: C9H8O4
Formula Weight: 180.16
Melting Point: 193-194°
Beilstein Reference: 2048750
MDL No.: MFCD00029972
Formel:
C₉H₈O₄ Molecular Weight: 180.16 g/mol Schmelzpunkt: 193…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00029972 CAS-Nummer: 1877-71-0 |