Benzoyl-DL-alanine ≥97%
Lieferant: Thermo Scientific
L13100.09
L13100.18
L13100.09EA
32.1
EUR
L13100.09
L13100.18
Benzoyl-DL-alanine ≥97%
Benzoyl-DL-alanine
Purity: 97+%
CAS No.: 1205-02-3
Molecular Formula: C10H11NO3
Formula Weight: 193.20
Melting Point: 163-167°
Beilstein Reference: 3201778
MDL No.: MFCD00020393
Formel:
C₁₀H₁₁NO₃ Molecular Weight: 193,20 g/mol Schmelzpunkt: 163…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020393 CAS-Nummer: 1205-02-3 EINECS: 214-879-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...