Ethyl mesitylglyoxylate 97%
Lieferant: Thermo Scientific
L13106.06
L13106.14
L13106.06EA
39.7
EUR
L13106.06
L13106.14
Ethyl mesitylglyoxylate 97%
Ethyl mesitylglyoxylate
Purity: 97%
CAS No.: 5524-57-2
Molecular Formula: C13H16O3
Formula Weight: 220.27
Refractive Index: 1.5120
Boiling Point: 164-165°/10mm
Beilstein Reference: 2970031
MDL No.: MFCD00151821
Formel:
C₁₃H₁₆O₃ Molecular Weight: 220,27 g/mol Siedepunkt: 164…165 °C (10 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151821 CAS-Nummer: 5524-57-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...