1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture) 96%
Lieferant: Thermo Scientific
Gefahr
Synonyme: 1,4-Di(aminomethyl)cyclohexane, Cyclohexane-1,4-diyldimethanamine, Hexahydro-p-xylylenediamine, 1,4-Cyclohexanebis(methylamine)
L13407.03 L13407.06 L13407.14
L13407.03EA 23.6 EUR
L13407.03 L13407.06 L13407.14
1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture) 96%
1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture)
Purity: 96% CAS No.: 2549-93-1 Molecular Formula: C8H18N2 Formula Weight: 142.25 Flash Point: 106°(222°F) Refractive Index: 1.4900 Boiling Point: 239-244° Beilstein Reference: 2071383 UN No.: UN2922 MDL No.: MFCD00039463
Formel: C₈H₁₈N₂
Molecular Weight: 142,25 g/mol
Siedepunkt: 239…244 °C
Dichte: 0,950
Flammpunkt: 106 °C (222 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00039463
CAS-Nummer: 2549-93-1
EINECS: 219-840-6
ADR: 8,III