5(6)-Carboxyfluorescein Gemisch aus Isomeren 97%
Lieferant: Thermo Scientific
Synonyme:
5(6)-FAM, 3′,6′-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-ar-carboxylic acid
L13439.03
L13439.MD
L13439.03EA
108
EUR
L13439.03
L13439.MD
5(6)-Carboxyfluorescein Gemisch aus Isomeren 97%
5(6)-Carboxyfluorescein
Purity: 97%
CAS No.: 72088-94-9
Molecular Formula: C21H12O7
Formula Weight: 376.32
Melting Point: ca 300°
MDL No.: MFCD00151081
Formel:
C₂₁H₁₂O₇ Molecular Weight: 752,63 g/mol Schmelzpunkt: ∼ 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151081 CAS-Nummer: 72088-94-9 EINECS: 276-331-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...