4-Chloro-2-fluorobenzylamine hydrochloride 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4-Chloro-2-fluorophenyl)methanamine hydrochloride

L13488.03 L13488.06
L13488.03EA 22.6 EUR
L13488.03 L13488.06
4-Chloro-2-fluorobenzylamine hydrochloride 97%
4-Chloro-2-fluorobenzylamine hydrochloride

Purity: 97% CAS No.: 202982-63-6 Molecular Formula: C7H7ClFN.HCl Formula Weight: 196.05 Melting Point: 243-245° MDL No.: MFCD00143286

Formel: C₇H₇ClFN·HCl
Molecular Weight: 196.05 g/mol
Schmelzpunkt: 243…245 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143286
CAS-Nummer: 202982-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR