3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propansäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Phthalimidopropionic acid, 3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

L13535.03 L13535.06
L13535.03EA 28.1 EUR
L13535.03 L13535.06
3-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)propansäure 98%
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid

Purity: 98% CAS No.: 3339-73-9 Molecular Formula: C11H9NO4 Formula Weight: 219.20 Melting Point: 151-153° Beilstein Reference: 15570 MDL No.: MFCD00023096

Formel: C₁₁H₉NO₄
Molecular Weight: 219,196 g/mol
Schmelzpunkt: 151…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023096
CAS-Nummer: 3339-73-9
EINECS: 222-083-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR