5-Bromo-3-pyridineacetic acid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromo-3-pyridylacetic acid

L13579.06 L13579.03
L13579.06EA 411 EUR
L13579.06 L13579.03
5-Bromo-3-pyridineacetic acid ≥98%
5-Bromo-3-pyridineacetic acid

Purity: 98+% CAS No.: 39891-12-8 Molecular Formula: C7H6BrNO2 Formula Weight: 216.03 Melting Point: 182-185° Beilstein Reference: 388264 MDL No.: MFCD00829308

Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
Schmelzpunkt: 182…185 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00829308
CAS-Nummer: 39891-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR