2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulphonyl chloride 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-(2,2,2-Trifluoroacetyl)-1,2,3,4-tetrahydroisoquinoline-7-sulphonyl chloride

L13587.03 L13587.06
L13587.03EA 38.5 EUR
L13587.03 L13587.06
2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulphonyl chloride 99%
2-Trifluoroacetyl-1,2,3,4-tetrahydroisoquinoline-7-sulphonyl chloride

Purity: 99% CAS No.: 74291-57-9 Molecular Formula: C11H9ClF3NO3S Formula Weight: 327.71 Melting Point: 102-106° Beilstein Reference: 5603124 UN No.: UN3261 MDL No.: MFCD01075651

Formel: C₁₁H₉ClF₃NO₃S
Schmelzpunkt: 102…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075651
CAS-Nummer: 74291-57-9
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR