2-Amino-4-(p-tolyl)thiazole 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-(p-Tolyl)-2-thiazolamine, 4-(4-Methylphenyl)-2-thiazolamine, 4-(p-Tolyl)thiazol-2-amine, 2-Amino-4-(4-methylphenyl)thiazole

L13863.03 L13863.06
L13863.03EA 26.7 EUR
L13863.03 L13863.06
2-Amino-4-(p-tolyl)thiazole 98%
2-Amino-4-(p-tolyl)thiazole

Purity: 98% CAS No.: 2103-91-5 Molecular Formula: C10H10N2S Formula Weight: 190.27 Melting Point: 134-138° Beilstein Reference: 143563 MDL No.: MFCD00170264

Formel: C₁₀H₁₀N₂S
Molecular Weight: 190,26 g/mol
Schmelzpunkt: 134…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00170264
CAS-Nummer: 2103-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR