N-Acetylneuraminic acid 97%

Lieferant: Thermo Scientific

Synonyme: Neu5Ac, (4S,5R,6R,7S,8R)-5-Acetamido-4,6,7,8,9-pentahydroxy-2-oxononanoic acid, NANA, Sialic acid, 5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic acid, Lactaminic acid

L13950.MB L13950.MC
L13950.MBEA 36.5 EUR
L13950.MB L13950.MC
N-Acetylneuraminic acid 97%
N-Acetylneuraminic acid

Purity: 97% CAS No.: 131-48-6 Molecular Formula: C11H19NO9 Formula Weight: 309.28 Storage Temperature: -30°C to -10°C Melting Point: ca 185° dec. Beilstein Reference: 1716283 Optical Rotation: -32° (c=0.5 in water) MDL No.: MFCD00006620

Formel: C₁₁H₁₉NO₉
Molecular Weight: 309,27 g/mol
Schmelzpunkt: ∼ 185 °C (Zers.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00006620
CAS-Nummer: 131-48-6
EINECS: 205-023-1
Merck Index: 14,8484

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR