3-(3-Pyridinyl)propansäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L14007.06
L14007.14
L14007.06EA
70.8
EUR
L14007.06
L14007.14
3-(3-Pyridinyl)propansäure 97%
3-(3-Pyridyl)propionic acid
Purity: 97%
CAS No.: 3724-19-4
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 156-159°
Beilstein Reference: 122232
MDL No.: MFCD00274197
Formel:
C₈H₉NO₂ Schmelzpunkt: 156…159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00274197 CAS-Nummer: 3724-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...